Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Leptin, Mouse, Recombinant Protein, E. coli
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
Recombinant, mouse leptin espressed in E. coli. Native leptin is a product of the obese (ob) gene that serves as a ligand for the OB receptor (OB-R). Mice with mutations in the ob gene have been found to be obese and diabetic and to have reduced activity, metabolism and body temperature. Suppresses insulin secretion by inhibiting activities of Ca2+-dependent PKC isoforms. Reported to reduce hepatic glucose production by blocking phosphoenolpyruvate synthesis.
Specifications
Specifications
| Formulation | Lyophilized from a sterile filtered PBS. |
| Preparation Method | Reconstitute with 20mM Tris-HCl, pH 8.0 to a final concentration of 1mg/mL. |
| Purification Method | SDS-PAGE |
| Quantity | 5 mg |
| Source | E. coli |
| Immunogen | Leptin |
| Storage Requirements | Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C or −70°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C or −70°C. Avoid freeze/thaw cycles of solutions. |
| Regulatory Status | RUO |
| Species | Mouse |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction