Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LiChroCART™ Columns/Purospher™ STAR Si Sorbents HPLC Cartridges, 5μm

Catalog No. M1502680001 Shop All MilliporeSigma Products
Change view
Click to view available options
Length (Metric):
125 mm
150 mm
250 mm
Diameter (Metric):
4 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
M1502680001 125 mm 4 mm
M1503560001 150 mm 4.6 mm
M1503570001 250 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1502680001 Supplier MilliporeSigma™ Supplier No. 1502680001
Add to Cart
Add to Cart

Offers exceptional separation efficiency for normal-phase chromatography of low molecular weight compounds soluble in organic solvents

  • Very high separation efficiency as measured by the plate count
  • Absence of metal impurities, thus giving consistently symmetrical peaks
  • Extended column lifetime

TRUSTED_SUSTAINABILITY

Specifications

Column Format Cartridge
Diameter (Metric) 4 mm
Length (Metric) 125 mm
Max. Pressure 2175 psi (150 bar) to 2900 psi (200 bar)
Particle Size 5 μm
pH 2 to 7.5
Pore Size 120 Å
Quantity 1/Pk.
Surface Area 330 m²/g
USP Type L3
Stationary Phase Silica
Packing Material Silica
Column Type Normal Phase
Phase Normal Phase
Product Type Cartridge
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.