Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MAP Kinase 2, Mouse, Recombinant Protein, E. coli

Catalog No. 4548502000U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-485-02000U 2000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-485-02000U Supplier MilliporeSigma™ Supplier No. 4548502000U
Add to Cart
Add to Cart
Recombinant, mouse MAP Kinase 2 expressed in E. coli. A serine/threonine protein kinase that acts at the end of a protein kinase cascade linking signals from cell surface receptor tyrosine kinases to cytoplasmic and nuclear events. This protein was co-expressed with MEK and is phosphorylated and active.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 50mM HEPES, 1mM DTT, 100μM Na2-EDTA, 50% glycerol, 0.01% BRIJ™ 35 detergent, pH 7.5.
Purification Method SDS-PAGE
Quantity 2000 U
Source E. coli
Immunogen MAP Kinase 2
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.