Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ pH Indicator Litmus Papers

Catalog No. M1094860003
Click to view available options
Color Indicator:
Blue

Includes

Color scale

Measure pH swiftly without using instruments

Suited for all media in environmental analysis and industrial in-process controls.

  • More precise evaluation due to differentiated color graduations
  • Biologically degradable polyester foil and low reagent content in reaction zones support problem-free waste disposal

  • Range: pH <7, red; pH >7, blue
  • M1094860003: blue
  • M1095180003: neutral
  • M1094890003: red
  • M1094860003 and M1094890003 are Reag. Ph. Eur grade

Order Info

Three rolls of 4.8m

TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Blue
For Use With (Application) For long term storage, since they are protected against external influences, pH indicator papers are manufactured by impregnating high quality filter paper with indicator or mixed indicator solutions
Includes With color scale
Quantity 3 Rolls
Storage Requirements 10°C to 25°C
Type Litmus Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.