Learn More
MilliporeSigma™ Remote Pure Water Dispenser
Catalog No. p-4908418 Shop All MilliporeSigma Products
Click to view available options
Model:
E-POD Pure Water Remote Dispenser
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Remote Pure Water Dispenser is compatible with E-POD and Q-POD systems.
Use for handheld glassware washing or similar tasks.
Certifications
ISO 9001 v.2000 and ISO 14001
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 v.2000, ISO 14001
Flow Rate 2 L/min.
Product Water Type Type 1
Resistivity 18.2MOhm
For Use With (Application) Provides Pressurized water at 2L/min.
For Use With (Equipment) Elix Advantage and Milli-Q Integral systems
Model E-POD Pure Water Remote Dispenser
Electrical Requirements 100/230 V 50/60 Hz
Voltage 100/230 V
Frequency 50/60 Hz
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.