Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Millipak™ Disposable Filter Units
Click to view available options
Connectors:
NPTM (In), SHB with Bell (Out)
SHB (In), SHB with Bell (Out)
Sanitary Flange (In/Out)
Flow Rate:
0.7 L/min. at 1.75 bar
1.1 L/min. at 1.75 bar
1.5 L/min. at 1.75 bar
1.9 L/min. at 1.75 bar
10.5 L/min. at 2.45 bar
2.8 L/min. at 1.75 bar
3.8 L/min. at 1.75 bar
6.4 L/min. at 2.45 bar
7.6 L/min. at 2 bar
7.6 L/min. at 700 mbar
8.5 L/min. at 2.45 bar
Volume (Metric):
10 L
20 L
30 L
Description
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.
- Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
- Point-of-use filter for high-purity water
- Ideally suited for high value-added applications
- Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
- Presterilized units are gamma irradiated
- Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
- 100% integrity tested to guarantee integrity

Specifications
Specifications
Connectors | SHB (In), SHB with Bell (Out) |
Disposable | Single-use |
Material | Polycarbonate |
Material (Membrane) | PVDF |
Pore Size | 0.45 μm |
Sterility | Sterile |
Filtration Area | 300 cm2 |
Flow Rate | 10.5 L/min. at 2.45 bar |
For Use With (Application) | Removal of particles and microorganisms from liquids and gases |
Inlet Connection (English) | 0.25 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction