Learn More
MilliporeSigma™ Mollifex™ Tissue Softener
Click to view available options
Quantity:
500 mL
Description
- Softening agent for use with paraffin embedded tissues
- Ideal for hard to section blocks and brittle tissues
Specifications
Specifications
Type | Mollifex Tissue Softener |
For Use With (Application) | Ideal for hard to section blocks and brittle tissues |
Quantity | 500 mL |