Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Ascorbic Acid Test Strips

Catalog No. m1100230001 Shop All MilliporeSigma Products
Click to view available options
Range:
50 to 2000mg/L

Includes

Tube containing 100 strips

To measure the levels of ascorbic acid in food

  • Small and easy to handle
  • Flexible and universal application
  • Phosphormolybdenum blue method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
  • Color-scale graduation: 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000mg/L

Reclose the tube containing the test strips immediately after use.

For food and water analysis

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Ideal for quickly measuring levels of natural ascorbic acid in foods such as fruit and vegetable juices, soft drinks
For Use With (Equipment) Colorimeter
Graduations 50-100-200-300-500-700-1000-2000mg/L
No. of Tests 100
Range 50 to 2000mg/L
Storage Requirements 2°C to 8°C
Test Method Phosphormolybdenum Blue
Type Test Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.