Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MQuant™ Peracetic Acid Test Strips
Click to view available options
Range:
100 to 500mg/L
5 to 50mg/L
500 to 2000mg/L
Description
Includes
Tube containing 100 strips
- Small and easy to handle
- Flexible and universal application
- Redox reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
1.10001.0001
Color-scale graduation: 100 - 150 - 200 - 250 - 300 - 400 - 500mg/L peracetic acid
1.10084.0001
Color-scale graduation: 5 - 10 - 20 - 30 - 50mg/L peracetic acid
Reclose the tube containing the test strips immediately after use.
For food and water analysis

Specifications
Specifications
For Use With (Application) | For food and water analysis |
For Use With (Equipment) | Colorimeter |
Graduations | 100-150-200-250-300-400-500mg/L |
No. of Tests | 100 |
Range | 100 to 500mg/L |
Storage Requirements | 2°C to 8°C |
Test Method | Redox Reaction |
Type | Test Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction