Learn More
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Shop All MilliporeSigma Products

Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | Standard |
| Lid | Yes |
| Membrane | Hydrophobic PVDF |
| Array | 8 x 12 |
| Pore Size | 0.45 μm |
| Product Line | MultiScreen |
| Color | White |
| Product Type | MultiScreen IP |
| Material | Acrylic |
| No. of Wells | 96 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.