Learn More
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Sample Preparation

Description
Formats are suitable for removal of large particulates, resin-based separations, solvent applications, and protein/DNA concentration or purification. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
- Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
- Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold
Choice of filter materials
- For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
- For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
- For assays that require solvent resistance, use plates with hydrophilic PTFE membranes
Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.
†MultiScreenHTS version features a recessed underdrain and rigid sidewalls.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Format | Standard |
Membrane | Hydrophilic PVDF |
Array | 8 x 12 |
Pore Size | 1.2 μm |
Color | Clear |
Product Type | MultiScreen-BV Plate |
Material | Styrene |
Volume (Metric) Well | 0.05 to 0.25 mL |
No. of Wells | 96 |
Sterility | Non-sterile |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.