Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ Column Loaders

Catalog No. p-155390 Shop All MilliporeSigma Products
Click to view available options
Volume (Metric) Well:
45 μL

No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly

  • Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Color Black
Volume (Metric) Well 45 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Load 45μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography
Quantity 1/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.