Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates

Catalog No. MSIPN4510 Shop All MilliporeSigma Products
Click to view available options
Pore Size:
0.45 μm

Non-sterile, clear 96-well filter plate with 0.45μm pore size Hydrophobic PVDF membrane for protein interaction assays, Avidin-biotin Linkages, DNA-binding proteins.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Format Standard
Membrane Hydrophobic PVDF
Array 8 x 12
Pore Size 0.45 μm
Binding Type Low Binding
Color Clear
Material Acrylic
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28 cm2
For Use With (Application) Protein interaction assays, avidin biotin linkages, DNA binding proteins
Quantity 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.