Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS Solubility Filter Plate

Catalog No. MSSLBPC50 Shop All MilliporeSigma Products
Click to view available options
Quantity:
10/Pk.
50/Pk.

Catalog No. MSSLBPC50

Supplier: MilliporeSigma™ MSSLBPC50

Only null left
Add to Cart
Add to Cart

96-well filter plate designed and optimized for aqueous solubility assays

  • High-throughput system to classify or quantify the aqueous solubility of compound libraries stored in solvent as a concentrated solution
  • Analysis time in under four hours using <100μg of sample per test
  • Automation compatible, with space for bar coding
  • Validated for high drug recovery
  • Low-binding membrane and low-binding plate materials
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Polycarbonate
Array 8 x 12
Pore Size 0.4 μm
Color Clear
Product Type MultiScreenHTS-PCF Filter Plate
Material Styrene
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Aqueous solubility assays
Quantity 50/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.