Learn More
MilliporeSigma™ Novagen™ NovaBlue GigaSingles™ Competent Cells
Catalog No. 712273 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
11 Reactions
22 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-227-3 11 Reactions
71-227-4 22 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-227-3 Supplier MilliporeSigma™ Supplier No. 712273
Add to Cart
Add to Cart
NovaBlue GigaSingles™ Competent Cells produce greater than 1 x 10⁹ colonies/μg plasmid DNA for cloning applications requiring high-efficiency transformations.
NovaBlue competent cells are for routine molecular cloning applications. NovaBlue is a K-12 strain derivative that offers high transformation efficiency, facilitates plasmid stability, and allows blue/white screening with appropriate plasmids.
TRUSTED_SUSTAINABILITY
Specifications
Efficiency Guaranteed efficiency: >1 x 109 cfu/μg
Content And Storage ≤-70°C
Quantity 11 Reactions