Learn More
Description
- Purified antibody conjugate produces a strong signal at a 1:4000 dilution for Western blots and dot blots
- Strep·Tag II Antibody HRP Conjugate can detect as little as 5ng Strep·Tag II fusion protein
- Formulation: Suspension in PBS, 5mg/mL BSA, 0.01% thimerosal, and stabilizing agents
Specifications
Specifications
| Product Type | Strep-Tag II Antibody HRP Conjugate |
| Quantity | 75 μL |
| Content And Storage | 2°C to 8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.