Learn More
MilliporeSigma™ Novagen™ Veggie BL21 (DE3) Singles™ Competent Cells
Catalog No. 712523 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
11 Reactions
22 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-252-3 11 Reactions
71-252-4 22 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-252-3 Supplier MilliporeSigma™ Supplier No. 712523
Add to Cart
Add to Cart
Protease-deficient hosts (BL21) for protein expression; Manufactured free of animal-derived media and components
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ≤-70°C
Quantity 11 Reactions