Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Pellicon™ XL Devices

Catalog No. p-117433
Click to view available options
MWCO:
10 kDa
100 kDa
1000 kDa
30 kDa
300 kDa
5 kDa
50 kDa
500 kDa
pH Range:
1 to 14
2 to 21

Includes

Operating instructions and Certificate of Quality

Small-volume tangential flow filter device

Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.

  • True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
  • Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
  • Controlled channel height
  • Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes

Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.

Order Info

Packaged one per box.

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
Material Polypropylene
MWCO 5 kDa
Pore Size 100 μm
Filtration Area 50 cm2
For Use With (Application) Protein purification, analytical sample preparation
Includes Operating instructions and certificate of quality
Max. Inlet Pressure 80 psi at 50°C
pH Range 1 to 14
Quantity 1 Pack
Recirculation Rate 0.03 to 0.05L/min.
Type Pellicon XL Ultrafiltration Device
Volume (English) Working 0.5 oz.
Volume (Metric) Hold Up 3.2 mL
Volume (Metric) Working 15 mL
Dimensions (L x W) 7.4 x 1.2 in. (18.8 x 3cm)
Length (English) 7.4 in.
Length (Metric) 18.8 cm
Width (English) 1.2 in.
Width (Metric) 3 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.