Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Plk2, GST- Fusion, Human, Recombinant Protein, S. frugiperda

Catalog No. 34610410UG
Click to view available options
Quantity:
10 μg
Full length, recombinant, human Plk2 fused at the N-terminus to GST and expressed in S. frugiperda insect cells using a baculovirus expression system. Plk2 is a G1 specific kinase that is specifically detected in brain, heart, and lung. Plk2 expression increases in the presence of DNA-damaging agents. The active kinase is useful for the study of Plk2 regulation and inhibitor screening.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 25mM Tris-HCl, 3mM DTT, 50% glycerol, 0.05% Tween™-20 detergent, pH 8.0.
Purification Method SDS-PAGE
Quantity 10 μg
Source S. frugiperda
Immunogen Plk2
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate GST-Fusion
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥70%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.