Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prothrombin, Human Plasma, Protein

Catalog No. p-5365265 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
2 mg
Native prothrombin from human plasma. Vitamin K-dependent glycoprotein synthesized in the liver. Conversion to thrombin is a key step in the blood coagulation pathway and catalyzes the coagulation of fibrinogen. Clinically, cases of selective deficiency are rare, although, in cases of liver cirrhosis, prothrombin is decreased.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM NaCl, 20mM Tris-HCl, pH 7.4.
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Prothrombin
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 3 months at −70°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.