Learn More
MilliporeSigma™ pT7Blue Blunt Vector (linearized)
Catalog No. 701744
Click to view available options
Quantity:
20 Reactions
40 Reactions
pT7Blue contains the pUC19 backbone (including high-copy number origin of replication and lac sequences), a T7 promoter, f1 origin of replication, and modified multiple cloning region. The multiple cloning region contains an EcoR V site used for T-cloning flanked by an Nde I site, which allows inserts to be conveniently subcloned into the Nde I site of many pET vectors.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 40 Reactions
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.