Learn More
MilliporeSigma™ pT7Blue Perfectly Blunt Cloning Kit
Catalog No. 701894 Shop All MilliporeSigma Products
Click to view available options
Quantity:
20 Reactions
40 Reactions
The pT7Blue-3 vector is multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, a T7 promoter and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned at each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 40 Reactions
Vector pT7Blue Perfectly Blunt Vector
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.