Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PureProteome™ Nickel Magnetic Beads

Catalog No. p-3607170 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
2 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGH02 2 x 1 mL
LSKMAGH10 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LSKMAGH02 Supplier MilliporeSigma™ Supplier No. LSKMAGH02
Only null left
Add to Cart
Add to Cart

Offer yields comparable with other magnetic purification systems while offering high purities

  • No lysate clearing—crude cell lysates are processed by magnetic beads
  • Fast processing time—entire protocol is completed in less than one hour
  • High capacity—bind 5 to 15mg of protein per mL of settled beads
  • High affinity—binding can take place in buffer containing <5mM EDTA
  • Flexible scale—varying the bead quantity allows different protein yields
  • Flexible elution—native or denatured proteins
  • Automatable—hands-free screening process

TRUSTED_SUSTAINABILITY

Specifications

Binding Property 1 to 5.5mg/mL
Content And Storage 2°C to 8°C
Particle Size 10 μm
Quantity 2 x 1 mL
For Use With (Application) Purification of His-tagged recombinant Protein
Type Magnetic Affinity Bead
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.