Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Nitrite Test Strips

Catalog No. p-4628040 Shop All MilliporeSigma Products
Change view
Click to view available options
Range:
0.5 to 25mg/L
Test Method:
Griess' reaction
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Test Method
M1169730001 0.5 to 25mg/L Griess' reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169730001 Supplier MilliporeSigma™ Supplier No. 1.16973.0001
Only null left
Add to Cart
Add to Cart

Includes

Tube containing 50 test strips, 1 bar-code strip.

To determine the presence or absence of residues of nitrite

  • Griess's reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Ideal for monitoring nitrite levels in drinking and groundwater as well as in-process, wastewater or aquariums, food and water analysis
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
Graduations 0.5 to 25mg/L
No. of Tests 50
Range 0.5 to 25mg/L
Storage Requirements 2°C to 8°C
Test Method Griess' reaction
Type Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.