Learn More
MilliporeSigma™ Replacement Parts for Stainless-Steel 13 and 25mm Filter Holders
Shop All MilliporeSigma Products
Click to view available options
Type:
Flat Gasket
Replacement Parts Kit
Support Screen
Description
Swinnex™ parts kit contains two PTFE gaskets, two O-rings, and two support screens.
Specifications
Specifications
| Material | PTFE |
| Type | Flat Gasket |