Learn More
Description
Stains Gram-negative bacteria red
Specifications
Specifications
| Type | Safranin |
| Quantity | 4000 mL |
| Format | Bottle |
| For Use With (Application) | Stains gram-negative bacteria red |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.