Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Silica Gel 60 Standardized Sorbents

Catalog No. p-4701052 Shop All MilliporeSigma Products
Click to view available options
Material:
Silica Gel G
Silica Gel H
Type:
Metal Can
Plastic Bottle
Plastic Drum
Quantity:
1 kg
2.5 kg
5 kg
25 kg

Catalog No. M1077369025

Supplier: MilliporeSigma™ 1077369025

Only null left
Add to Cart
Add to Cart

Silica gel 60 for thin-layer chromatography. Silica gel 60 G CAS No. 7631-86-9, EC Number 231-545-4.

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40μm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 to 40 μm
Grade TLC
Packaging Steel Drum
pH 7
Quantity 25 kg
Type Plastic Drum
Material Silica Gel H
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.