Learn More
MilliporeSigma™ Silica Gel 60G TLC Plates, Glass Backed
Catalog No. p-4833037
Click to view available options
Type:
TLC Silica Gel 60 G
TLC Silica Gel 60 G F254
These new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph Eur monograph methods, which require TLC plates with gypsum binder.
  • 20 x 20cm
  • Provided in packs of 25
  • For thin layer chromatography applications
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25/Pk.
Type TLC Silica Gel 60 G
Dimensions (L x W) 20 x 20 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.