Learn More
MilliporeSigma™ SNAP i.d.™ Protein Detection System: Blot Holders
Catalog No. WBAVDBH011 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Double Well Blot Holder
Single Well Blot Holder
Quantity:
60/Pk.
Device Holds:
4.6 x 8.8 cm Blots
7.5 x 8.8 cm Blots
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity Device Holds
WBAVDBH011 Single Well Blot Holder 60/Pk. 7.5 x 8.8 cm Blots
WBAVDBH022 Double Well Blot Holder 60/Pk. 4.6 x 8.8 cm Blots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. WBAVDBH011 Supplier MilliporeSigma™ Supplier No. WBAVDBH011
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Blot holders for use with SNAP i.d. Protein Detection System
TRUSTED_SUSTAINABILITY
Specifications
Device Holds 7.5 x 8.8 cm Blots
For Use With (Application) Western Blot
For Use With (Equipment) Snap i.d.™ Protein Detection System
Type Single Well Blot Holder
Includes Spacers
No. of Wells 1
Quantity 60/Pk.