Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Synergy™ Ultrapure Water Purification System
GSA_VA

Catalog No. p-4426713
Click to view available options
Total Organic Carbon:
<5 ppb
Includes:
Remote Dispenser, 185nm Photooxidation UV Lamp

Obtain Type 1 reagent grade water with a flow rate above 1L/min when using the Millipore Synergy™ Ultrapure Water Purification System.

Synergy systems are designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water.

  • Produce Type 1 water (18.2 MOhm. cm at 25°C ultrapure water) on demand from pretreated water (reverse osmosis or demineralized water)
  • Maintenance reduced to a single cartridge change once or twice a year, depending on the volume produced and the quality of the pure water fee
  • Designed for users needing between 2 and 100 liters of ultrapure water per day
  • Quality suitable for HPLC and other spectrophotometric/spectroscopic analytical techniques for biological, biochemical, and cell culture applications
  • Dispensing adapted to different size laboratory glassware
  • Time-saving fixed-volume dispense
  • Compact design, along with the versatile Remote dispenser, ensure easy installation anywhere in your laboratory

  • Dimensions: 15D x 21.3W x 11.4 in. H (38 x 54 x 29cm)
  • Electrical feed voltage: 100-250V ±10%
  • Electrical feed frequency:50-60Hz ±10%

  • Synergy UV System: For applications requiring very low organic contaminant levels,it incorporates a dual wavelength UV lamp to produce water with <5ppb TOC (when feed water TOC is below 50ppb), making it suitable for HPLC, GC, ILC and TOC analyses

LC buffer preparation, HPLC isocratic and gradient solution preparation, Sample dilution, Buffer reagents for molecular biology and cell culture media preparation, Preparation of chemical solutions used with titrators, spectrophotometers and electrophoresis systems

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Feedwater Source Pretreated
Flow Rate 1 L/min.
Product Water Type Type 1
Resistivity 18.2 Mohm.cm product water
Total Organic Carbon <5 ppb
Type UV Pure to Pure and Ultrapure Purification System
Feedwater Pressure 4.3 psi (0.3 bar)
For Use With (Application) Designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water
Includes Remote Dispenser, 185nm Photooxidation UV Lamp
Width (English) 11.4 in.
Width (Metric) 29 cm
Height (English) 21.3 in.
Height (Metric) 54 cm
Depth (English) 15 in.
Depth (Metric) 38 cm
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.