Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Thrombin, Human Plasma, Protein

Catalog No. p-5365300
Click to view available options
Quantity:
100 U
1000 U
Key enzyme that activates factor XIII and converts fibrinogen into fibrin.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 200mM NaCl, 50mM citrate buffer, 0.1% PEG-8000, pH 6.5. Contains BSA as a stabilizer.
Preparation Method Aqueous Buffers or H2O (1mg/mL)
Purification Method Complete activation from homogeneous prothrombin by SDS-PAGE
Quantity 100 U
Immunogen Thrombin
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 2 months at −70°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.