Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Transferrin, Apo-, Low Endotoxin Grade, Human Plasma, Protein

Catalog No. 1784811GM
Click to view available options
Quantity:
100 mg
1 g
Native apotransferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free. Cell culture-tested.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 10mM phosphate buffer, pH 7.4.
Preparation Method H2O (10mg/mL)
Purification Method SDS-PAGE
Quantity 1 g
Immunogen Transferrin, Apo-
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 year at −20°C.
Regulatory Status RUO
Endotoxin Concentration Low Endotoxin Grade
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.