Learn More
MilliporeSigma™ Ultrafiltration Discs, 10 kDa NMW, Biomax Polyethersulfone
Shop All MilliporeSigma Products

Description
10 kDa NMWL, Biomax polyethersulfone. Assemble your Amicon Stirred Cell with the membrane of your choice.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.
Features & Benefits:
- Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
- Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements
- Compatible with Amicon™ Stirred Cells
Applications:
Concentration, Buffer Exchange, Desalting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Ultrafiltration Disc |
| Material | PES |
| MWCO | 10 kDa |
| Filtration Area | 13.4 cm2 |
| For Use With (Application) | Protein purification; protein concentration; protein desalting |
| Housing Material | PES |
| Quantity | 10Pack |
| Diameter (English) | 1.75 in. |
| Diameter (Metric) | 44.5 mm |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.