Learn More
MilliporeSigma™ Ultrafiltration Membranes - BioMax™ PB21
High-recovery membranes for stirred cells
Manufacturer:  MilliporeSigma™ PBQK07610
 View more versions of this product
Catalog No. PBQK07610
Add to cart
Description
Description
  • High-flow, polyethersulfone (PES) membranes
  • Use to concentrate faster or to concentrate and desalt more concentrated samples
  • Open microstructure provides extremely fast separation capability
  • Recommended for serum, plasma, or conditioned tissue culture media samples