Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Urokinase, Human Urine, Protein
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 kU
Description
Native urokinase from human urine. A thrombolytic agent that converts plasminogen to plasmin. A mixture of high molecular weight (54kDa) and low molecular weight (33kDa) urokinase.
Specifications
Specifications
| Formulation | Lyophilized from 200mM mannitol, 100mM NaCl, 50mM Tris-HCl, 0.1% PEG, pH 7.5. |
| Preparation Method | Reconstitute in distilled H2O |
| Quantity | 10 kU |
| Immunogen | Urokinase |
| Storage Requirements | Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 6 months at −70°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction