Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
GSA_VA
Catalog No. p-169856
Click to view available options
For Use With (Equipment):
A10 TOC Monitor (external), Milli-Q Ultrapure A10, High-volume Elix TOC Upgrade Water Purification Kit
ASM Pure Water, Older SDS Reservoir
Elix 20/35/70/100, RiOs 35/50/100/150/200
Elix 3/5/10
Milli-Q Advantage a10 TOC Monitor
Milli-Q gradient (purple door), Synthesis (orange door) Model
Synergy UV, Simplicity UV
Wavelength:
185 nm
254 nm
254/185 nm
Control bacteria and oxidize organics
  • Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type UV Lamp
For Use With (Equipment) Elix 3/5/10
Wavelength 254 nm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.