Learn More
MilliporeSigma™ Vasoactive Intestinal Peptide, Human, Porcine, and Rat, Calbiochem
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Species | Human, Porcine, Rat |
| Purification Method | HPLC |
| Purity | ≥95% |
| CAS | 40077-57-4 |
| Color | White to Off-white |
| Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. |
| Format | Lyophilized Solid |
| Formulation | Supplied as a trifluoroacetate salt. |
| Product Type | Vasoactive Intestinal Peptide |
| Quantity | 0.5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.