Learn More
MilliporeSigma™ Vent Filters for Pure Water Storage Reservoirs
Shop All MilliporeSigma Products
Description
For use with Pure Water Reservoirs
- Polyethylene vent filters protect purified water stored in Pure Water Reservoirs when used with low-volume Elix™ or RiOs™ Water Purification Systems from ingress of airborne microorganisms
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Material | Polyethylene |
For Use With (Equipment) | RiOs™ 5/8/16 system |
Hydrophobicity | Hydrophobic |
Type | Vent Filter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.