Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Xanthine Oxidase, Buttermilk, Protein

Catalog No. p-5365328 Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 U
50 U
Native xanthine oxidase from buttermilk. Complex metalloprotein that catalyzes oxidative hydroxylation of a variety of aromatic heterocycles and simple aldehydes. Catalyzes the oxidation of xanthine to uric acid.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation Crystalline suspension in 60% ammonium sulfate.
Quantity 10 U
Immunogen Xanthine Oxidase
Regulatory Status RUO
Species Bovine (Buttermilk)
Recombinant Native
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.