Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMD Mouse anti-Human, Unconjugated, Clone: EMD/2168, Abnova™

Catalog No. 89337423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-423 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-423 Supplier Abnova Corporation Supplier No. MAB20928
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

EMD Monoclonal antibody specifically detects EMD in Human samples. It is validated for IHC-P.

Specifications

Antigen EMD
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EMD/2168
Conjugate Unconjugated
Description emerin
Gene EMD
Gene Alias EDMD, LEMD5, STA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 56-167 of human EMD
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2010
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.