Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EME1 Antibody (MTA31 7h2/1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200058419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395444 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395444 Supplier Novus Biologicals Supplier No. NBP2503860.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EME1 Monoclonal antibody specifically detects EME1 in Human samples. It is validated for Western Blot

Specifications

Antigen EME1
Applications Western Blot
Classification Monoclonal
Clone MTA31 7h2/1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS
Gene Alias crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A
Host Species Mouse
Immunogen Human EME1
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 146956
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.