Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EME1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597028 Supplier Invitrogen™ Supplier No. PA597028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MALKKSSPSL DSGDSDSEEL PTFAFLKKEP SSTKRRQPER EEKIVVVDIS DCEASCPPAP ELFSPPVPEI AETVTQTQPV RLLSSESEDE EEFIPLAQRL TCKFLTHKQL SPEDSSSPVK SVLDHQNNEG ASCDWKKPFP KIPEVPLHDT PERSAADNKD LILDPCCQLP AYLSTCPGQS SSLAVTKTNS DILPPQKKTK PSQKVQGRGS HGCRQQRQAR QKESTLRRQE RKNAALVTRM KAQRPEECLK HIIVVLDPVL LQMEGGGQLL GALQTMECRC VIEAQAVPCS VTWRRRAGPS EDREDWVEEP TVLVLLRAEA; Positive Samples: Mouse thymus, Rat thymus; Cellular Location: Nucleus, nucleolus.

EME1 and MUS81 form an endonuclease complex that cleaves branched DNA structures, especially those arising during stalled DNA replication (Abraham et al., 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EME1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EME1
Gene Accession No. Q8BJW7
Gene Alias 6820428D13; Crossover junction endonuclease EME1; EME1; essential meiotic endonuclease 1 homolog 1; essential meiotic endonuclease 1 homolog 1 (S. pombe); essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease 1; hMMS4; homolog of yeast EME1 endonuclease; MMS4; MMS4 homolog; MMS4L; Msm4; SLX2 structure-specific endonuclease subunit homolog A; SLX2A
Gene Symbols EME1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human EME1 (NP_6896762).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268465, 287634
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.