Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EME1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA527110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527110 Supplier Thermo Scientific Supplier No. PA527110

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EME1 and MUS81 form an endonuclease complex that cleaves branched DNA structures, especially those arising during stalled DNA replication (Abraham et al., 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EME1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene EME1
Gene Accession No. Q96AY2
Gene Alias 6820428D13; Crossover junction endonuclease EME1; EME1; essential meiotic endonuclease 1 homolog 1; essential meiotic endonuclease 1 homolog 1 (S. pombe); essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease 1; hMMS4; homolog of yeast EME1 endonuclease; MMS4; MMS4 homolog; MMS4L; Msm4; SLX2 structure-specific endonuclease subunit homolog A; SLX2A
Gene Symbols EME1
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 1 and 252 of Human EME1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146956
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.