Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EME1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988077
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8077 Supplier Bioss Supplier No. BS7861R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EME1 and MUS81 form an endonuclease complex that cleaves branched DNA structures, especially those arising during stalled DNA replication (Abraham et al., 2003).

Specifications

Antigen EME1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EME1
Gene Accession No. Q96AY2
Gene Alias 6820428D13; Crossover junction endonuclease EME1; EME1; essential meiotic endonuclease 1 homolog 1; essential meiotic endonuclease 1 homolog 1 (S. pombe); essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease 1; hMMS4; homolog of yeast EME1 endonuclease; MMS4; MMS4 homolog; MMS4L; Msm4; SLX2 structure-specific endonuclease subunit homolog A; SLX2A
Gene Symbols EME1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EME1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146956
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.