Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EME2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561090
Description
Immunogen sequence: ALAQYPLKQY RESQAFSFCT AGRWAAGEPV ARDGAGLQAA WRRQIRQFSR VS Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
EME2 forms a heterodimer with MUS81 that functions as an XPF-type flap/fork endonuclease in DNA repair.
Specifications
EME2 | |
Polyclonal | |
Unconjugated | |
EME2 | |
EME2; essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease subunit 2; gs125; homolog of yeast EME1 endonuclease 2; probable crossover junction endonuclease EME2; SLX2 structure-specific endonuclease subunit homolog B; SLX2B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
197342 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A4GXA9 | |
EME2 | |
Recombinant protein corresponding to Human EME2. Recombinant protein control fragment (Product #RP-93647). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction