Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Emerin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1751R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1751R7 7 mL Prediluted
MS1751S0 100 μL Concentrated
MS1751S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1751R7 Supplier Epredia™ Supplier No. MS1751R7

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ Emerin Ab-1, Mouse Monoclonal Antibody.

Emerin is a serine-rich, ubiquitously expressed protein with the highest concentration in skeletal and cardiac muscles. It is localized in the nuclear membrane and is coded by the STA gene at the  Xq28 locus. Emerin deficiency is the main causal feature of Emery-Dreifuss muscular dystrophy (EDMD), which is an X-linked disorder leading to weakening of muscles and cardiomyopathy presented as heart block.

Host Species: Mouse

Clone: 4G5

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Near N-terminus

Immunogen: Recombinant protein corresponding to 222 amino acid fragment near N-terminus of human emerin protein

Molecular Weight: 34 kDa

Positive Control: Tonsil

Cellular Localization: Perinuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Emerin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4G5
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to 222 amino acid fragment near N-terminus of human emerin protein
Quantity 7 mL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.