Learn More
Description
Host Species: Mouse
Clone: 4G5
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Near N-terminus
Immunogen: Recombinant protein corresponding to 222 amino acid fragment near N-terminus of human emerin protein
Molecular Weight: 34 kDa
Positive Control: Tonsil
Cellular Localization: Perinuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Emerin Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 4G5 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to 222 amino acid fragment near N-terminus of human emerin protein |
| Purification Method | Tonsil |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.