Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Emerin Monoclonal Antibody (CL0201)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531327
Description
Immunogen sequence: ASSYSFSDLN STRGDADMYD LPKKEDALLY QSKGYNDDYY EESYFTTRTY GEPESAGPSR AVRQSVTSFP DADAFHHQVH DDDLLSSSEE ECKDRERPMY GRDSACQSIT HY.
Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
Specifications
Emerin | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P50402 | |
Emd | |
Recombinant protein corresponding to Human EMD. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
CL0201 | |
Unconjugated | |
Emd | |
AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA | |
Mouse | |
Protein A | |
RUO | |
2010 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction