Learn More
Invitrogen™ Emerin Monoclonal Antibody (MANEM1(5D10))

Description
This antibody does not cross-react with mouse samples. This target has a predicted molecular weight of 29 kDa.
Specifications
Specifications
| Antigen | Emerin |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | MANEM1(5D10) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Emd |
| Gene Accession No. | P50402 |
| Gene Alias | AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.