Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Emerin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Emerin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7857
![]() |
Bethyl Laboratories, Inc
A304491AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Emerin is a serine rich protein ubiquitously expressed with highest concentration in skeletal and cardiac muscles. It is localized in the nuclear membrane and is coded by STA gene at Xq28 locus. Emerin deficiency is tye main causal feature of Emery-Dreifuss muscular dystrophy (EDMD), which is a X-linked disorder leading to weakening of muscles and cardiomyopathy presented as heart block.Specifications
Emerin | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Emerin; STA; EDMD; EMD | |
EMD | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P50402 | |
2010 | |
Between 1 and 50 | |
Primary | |
4° C | |
EMD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title